Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286538.1 | NP_001273467.1 | 3 | 522 | ||
NM_001286540.1 | NP_001273469.1 | 3 | 522 | |||
NM_001286541.1 | NP_001273470.1 | 2 | 321 | |||
NM_001286551.1 | NP_001273480.1 | 3 | 475 | |||
NM_001286605.1 | NP_001273534.1 | 3 | 475 | |||
NM_011014.3 | NP_035144.1 | 3 | 522 | |||
GenBank mRNA | AF004927.1 | - | 3 | 476 | ||
AF030198.1 | - | 3 | 436 | |||
AK002718.1 | - | 3 | 455 | |||
AK154300.1 | - | 3 | 494 | |||
AK159886.1 | - | 3 | 456 | |||
AY390765.1 | - | 3 | 415 | |||
AY390766.1 | - | 3 | 415 | |||
AY390768.1 | - | 2 | 214 | |||
BC002000.1 | - | 3 | 471 | 3484605 | ||
BC019930.1 | - | 3 | 450 | 5123891 | ||
BM507398.1 | - | 3 | 367 |