Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286538.1 | NP_001273467.1 | 2, 3 | 467 | ||
NM_001286540.1 | NP_001273469.1 | 2, 3 | 467 | |||
NM_001286541.1 | NP_001273470.1 | 1, 2 | 266 | |||
NM_011014.3 | NP_035144.1 | 2, 3 | 467 | |||
GenBank mRNA | AF004927.1 | - | 2, 3 | 421 | ||
AF030198.1 | - | 2, 3 | 381 | |||
AK002718.1 | - | 2, 3 | 400 | |||
AK154300.1 | - | 2, 3 | 439 | |||
AK159886.1 | - | 2, 3 | 401 | |||
AY390765.1 | - | 2, 3 | 360 | |||
AY390766.1 | - | 2, 3 | 360 | |||
AY390768.1 | - | 1, 2 | 159 | |||
BB851853.1 | - | 2, 3 | 468 | |||
BC002000.1 | - | 2, 3 | 416 | 3484605 | ||
BC019930.1 | - | 2, 3 | 395 | 5123891 |