Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162387.1 | NP_001155859.1 | 3 | 539 | ||
NM_001162388.1 | NP_001155860.1 | 3 | 303 | |||
NM_011069.3 | NP_035199.3 | 3 | 539 | |||
GenBank mRNA | AF093671.1 | - | 3 | 284 | ||
AK005236.1 | - | 3 | 291 | |||
AK011709.1 | - | 3 | 337 | |||
AK082279.1 | - | 3 | 298 | |||
AK082778.1 | - | 3 | 260 | |||
BC013812.1 | - | 3 | 288 | 3483892 | ||
BC054414.1 | - | 3 | 287 | 3257217 | ||
BQ443231.1 | - | 3 | 349 | |||
BY285331.1 | - | 3 | 369 | |||
CD806660.1 | - | 3 | 529 |