Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN2 SIRNA Assays ›
          Gene Symbol:
          Ptpn2
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 2
          Gene Aliases:
          AI325124, Ptpt, TC-PTP
          Chromosome Location:
          Chr.18: 67665501 - 67724748 on Build GRCm38
          UniGene ID:
          Mm.260433
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s72452

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ptpn2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127177.1 NP_001120649.1 8 1039
          NM_008977.3 NP_033003.1 8 1039
          GenBank mRNA AK014528.1 - 8 1006
          AK076072.1 - 8 992
          AK132013.1 - 8 1002
          BC008269.1 - 8 1013 3602983
          M80739.1 - 8 994
          M81477.1 - 8 994
          S52655.1 - 8 989

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          negative regulation of cell proliferation
          insulin receptor signaling pathway
          negative regulation of tumor necrosis factor-mediated signaling pathway
          negative regulation of lipid storage
          dephosphorylation
          B cell differentiation
          T cell differentiation
          erythrocyte differentiation
          peptidyl-tyrosine dephosphorylation
          negative regulation of epidermal growth factor receptor signaling pathway
          negative regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of tyrosine phosphorylation of Stat6 protein
          glucose homeostasis
          negative regulation of macrophage differentiation
          positive regulation of gluconeogenesis
          negative regulation of insulin receptor signaling pathway
          negative regulation of inflammatory response
          negative regulation of T cell receptor signaling pathway
          negative regulation of chemotaxis
          negative regulation of interferon-gamma-mediated signaling pathway
          negative regulation of type I interferon-mediated signaling pathway
          negative regulation of protein tyrosine kinase activity
          negative regulation of interleukin-6-mediated signaling pathway
          negative regulation of ERK1 and ERK2 cascade
          regulation of hepatocyte growth factor receptor signaling pathway
          negative regulation of interleukin-2-mediated signaling pathway
          negative regulation of prolactin signaling pathway
          negative regulation of interleukin-4-mediated signaling pathway
          negative regulation of macrophage colony-stimulating factor signaling pathway
          negative regulation of positive thymic T cell selection
          positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of PERK-mediated unfolded protein response
          negative regulation of platelet-derived growth factor receptor-beta signaling pathway
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          integrin binding
          protein binding
          hydrolase activity
          phosphatase activity
          protein kinase binding
          syntaxin binding
          receptor tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0