Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL12RB1 SIRNA Assays ›
          Gene Symbol:
          IL12RB1
          Gene Name:
          interleukin 12 receptor subunit beta 1
          Gene Aliases:
          CD212, IL-12R-BETA1, IL12RB, IMD30
          Chromosome Location:
          Chr.19: 18058994 - 18099027 on Build GRCh38
          UniGene ID:
          Hs.567294
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7363

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL12RB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290023.1 NP_001276952.1 3 335
          NM_001290024.1 NP_001276953.1 4 326
          NM_005535.2 NP_005526.1 3 335
          NM_153701.2 NP_714912.1 3 335
          XM_006722741.3 XP_006722804.2 4 528
          XM_011527966.2 XP_011526268.1 4 549
          XM_011527967.2 XP_011526269.1 4 535
          XM_011527968.2 XP_011526270.1 4 543
          XM_011527969.2 XP_011526271.1 4 529
          XM_011527970.2 XP_011526272.1 4 548
          XM_011527971.2 XP_011526273.1 4 546
          XM_011527972.2 XP_011526274.1 4 546
          XM_011527973.2 XP_011526275.1 3 340
          XM_011527974.2 XP_011526276.1 3 328
          XM_011527975.2 XP_011526277.1 4 526
          XM_011527976.2 XP_011526278.1 4 546
          XM_011527977.2 XP_011526279.1 4 519
          GenBank mRNA AK225919.1 - 3 266
          AK290423.1 - 3 263
          BC029121.1 - 3 224
          BC137404.1 - 3 180 9021402
          BC137406.1 - 3 180 9021404
          BX647221.1 - 4 326
          DA008204.1 - 3 335
          U03187.1 - 3 219

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          positive regulation of T-helper 1 type immune response
          signal transduction
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          positive regulation of interferon-gamma production
          interleukin-12-mediated signaling pathway
          interleukin-23-mediated signaling pathway
          positive regulation of activated T cell proliferation
          positive regulation of memory T cell differentiation
          cellular response to interferon-gamma
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 17 cell lineage commitment
          cytokine receptor activity
          interleukin-12 receptor binding
          interleukin-12 receptor activity
          interleukin-23 binding
          interleukin-23 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix