Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034962.1 | NP_001030134.1 | 5 | 407 | ||
NM_001034963.1 | NP_001030135.1 | 4 | 317 | |||
NM_001034964.1 | NP_001030136.1 | 3 | 232 | |||
NM_009166.3 | NP_033192.2 | 6 | 596 | |||
NM_178362.1 | NP_848139.1 | 5 | 683 | |||
GenBank mRNA | AF078666.1 | - | 4 | 306 | ||
AF078667.1 | - | 5 | 396 | |||
AF521593.1 | - | 2 | 189 | |||
AK035212.1 | - | 5 | 399 | |||
AK087553.1 | - | 5 | 683 | |||
AK122487.2 | - | 5 | 351 | |||
AK146712.1 | - | 3 | 235 | |||
AK166943.1 | - | 4 | 374 | |||
AK208107.1 | - | 1 | 61 | |||
BC012703.1 | - | 5 | 396 | 3602997 | ||
U58883.1 | - | 6 | 534 |