Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253862.1 | NP_001240791.1 | 15 | 1575 | ||
NM_001253863.1 | NP_001240792.1 | 9 | 1026 | |||
NM_001253864.1 | NP_001240793.1 | 8 | 954 | |||
NM_001253865.1 | NP_001240794.1 | 10 | 1201 | |||
NM_011544.3 | NP_035674.2 | 16 | 1704 | |||
GenBank mRNA | AK132002.1 | - | 9 | 1014 | ||
AK133958.1 | - | 9 | 1014 | |||
AK134037.1 | - | 15 | 1576 | |||
AK135376.1 | - | 8 | 922 | |||
BC037097.1 | - | 10 | 1189 | 5345693 | ||
M97635.1 | - | 16 | 1675 | |||
M97636.1 | - | 1 | 1679 | |||
X64840.1 | - | 16 | 1504 |