Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vpreb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016983.1 | NP_058679.1 | 2 | 137 |
Gene Symbol: Vpreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016982.2 | NP_058678.1 | 2 | 281 | ||
GenBank mRNA | AK010843.1 | - | 2 | 275 | ||
AK010879.1 | - | 2 | 275 | |||
BC117051.1 | - | 2 | 243 | 40126370 | ||
BC119175.1 | - | 1 | 330 | 8733924 | ||
BC144850.1 | - | 1 | 330 | 9053398 | ||
X05556.1 | - | 2 | 276 |