Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK8 SIRNA Assays ›
          Gene Symbol:
          Mapk8
          Gene Name:
          mitogen-activated protein kinase 8
          Gene Aliases:
          AI849689, JNK, JNK1, Prkm8, SAPK1
          Chromosome Location:
          Chr.14: 33377898 - 33447214 on Build GRCm38
          UniGene ID:
          Mm.21495
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s77121

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mapk8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016700.4 NP_057909.1 11 1291
          GenBank mRNA AB005663.1 - 10 1124
          AB206575.1 - 10 1066
          AB219977.1 - 10 1100
          AK043102.1 - 11 1291
          AK139498.1 - 11 1279
          BC053027.1 - 11 1261 5694771

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          regulation of transcription, DNA-templated
          protein phosphorylation
          inflammatory response
          response to osmotic stress
          signal transduction
          JNK cascade
          JUN phosphorylation
          response to heat
          response to UV
          regulation of gene expression
          positive regulation of gene expression
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          positive regulation of cell migration
          regulation of histone deacetylation
          positive regulation of microtubule polymerization
          neuron projection development
          positive regulation of cyclase activity
          negative regulation of protein binding
          regulation of protein localization
          response to hydrogen peroxide
          regulation of circadian rhythm
          negative regulation of apoptotic process
          positive regulation of DNA replication
          response to cadmium ion
          determination of dorsal identity
          rhythmic process
          positive regulation of protein metabolic process
          cellular response to hydrogen peroxide
          cellular response to lipopolysaccharide
          cellular response to nitric oxide
          positive regulation of deacetylase activity
          type B pancreatic cell apoptotic process
          neuronal stem cell population maintenance
          programmed necrotic cell death
          positive regulation of determination of dorsal identity
          positive regulation of apoptotic signaling pathway
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          JUN kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          enzyme binding
          histone deacetylase regulator activity
          histone deacetylase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix