Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dysf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077694.2 | NP_001071162.1 | 48 | 5774 | ||
NM_021469.3 | NP_067444.2 | 48 | 5592 | |||
GenBank mRNA | AF188290.2 | - | 47 | 5367 | ||
AK087986.1 | - | 12 | 1464 | |||
AK131144.1 | - | 48 | 5589 | |||
BC043692.1 | - | 8 | 964 |