Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016805.2 | NP_058085.2 | 12 | 2324 | ||
GenBank mRNA | AF073992.1 | - | 12 | 2261 | ||
AK037517.1 | - | 12 | 2267 | |||
AK089058.1 | - | 12 | 2325 | |||
AK145499.1 | - | 12 | 2327 | |||
AK145682.1 | - | 12 | 2327 | |||
AK159084.1 | - | 12 | 2293 | |||
AK159614.1 | - | 12 | 2329 | |||
AK159956.1 | - | 12 | 2266 | |||
AK165844.1 | - | 12 | 2324 | |||
AK167200.1 | - | 12 | 2327 | |||
AK168657.1 | - | 12 | 2266 | |||
AK207780.1 | - | 3 | 357 | |||
BC018353.1 | - | 12 | 2169 | 3584692 |