Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161800.1 | NP_001155272.1 | 6 | 874 | ||
NM_026448.3 | NP_080724.2 | 6 | 874 | |||
GenBank mRNA | AK012326.1 | - | 6 | 855 | ||
AK083358.1 | - | 6 | 860 | |||
AK089072.1 | - | 6 | 859 | |||
BC029154.1 | - | 6 | 841 | 4952319 |