Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205253.1 | NP_001192182.1 | 4 | 1815 | ||
NM_001293688.1 | NP_001280617.1 | 4 | 1799 | |||
NM_019576.2 | NP_062522.1 | 5 | 1958 | |||
GenBank mRNA | AB039946.1 | - | 4 | 1786 | ||
AK014758.1 | - | 1 | 257 | |||
AK049636.1 | - | 5 | 1921 | |||
AK052555.1 | - | 4 | 1756 | |||
BC051957.1 | - | 4 | 1756 | |||
BC119603.2 | - | 4 | 1786 | 40060210 |