Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020003.1 | NP_064387.1 | 2 | 229 | ||
GenBank mRNA | AB027141.1 | - | 2 | 185 | ||
AB027142.1 | - | 2 | 229 | |||
AK002728.1 | - | 2 | 193 | |||
AK007803.1 | - | 2 | 202 | |||
AK075614.1 | - | 2 | 207 | |||
AK150794.1 | - | 2 | 202 | |||
AK154340.1 | - | 2 | 202 | |||
AK160793.1 | - | 169 | ||||
AK170696.1 | - | 2 | 192 | |||
AK171042.1 | - | 2 | 201 | |||
AK171142.1 | - | 675 | ||||
AK172606.1 | - | 1 | 668 | |||
BC021547.1 | - | 2 | 195 | 4507218 |