Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020618.4 | NP_065643.1 | 4 | 737 | ||
GenBank mRNA | AK018493.1 | - | 4 | 166 | ||
AK076177.1 | - | 4 | 169 | |||
BC034509.1 | - | 4 | 152 | |||
BC047141.1 | - | 4 | 158 | 6397290 | ||
BC061498.1 | - | 4 | 165 | 5251562 | ||
BC065043.1 | - | 4 | 737 | 5685483 | ||
EU327024.1 | - | 3 | 115 |