Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199275.2 | NP_001186204.1 | 6 | 740 | ||
NM_001199276.2 | NP_001186205.1 | 4 | 410 | |||
NM_001271455.1 | NP_001258384.1 | 5 | 463 | |||
NM_001271456.1 | NP_001258385.1 | 6 | 635 | |||
NM_021327.4 | NP_067302.2 | 5 | 582 | |||
GenBank mRNA | AJ242777.1 | - | 4 | 382 | ||
AJ242778.1 | - | 5 | 554 | |||
AK135338.1 | - | 5 | 561 | |||
AK156131.1 | - | 5 | 435 | |||
AK170841.1 | - | 4 | 388 | |||
AK172890.1 | - | 368 | ||||
AK199662.1 | - | 2 | 103 | |||
AK199664.1 | - | 2 | 103 | |||
AY012159.2 | - | 2 | 152 | |||
BC008186.1 | - | 5 | 499 | 3589311 | ||
BC008665.1 | - | 6 | 606 | 3598934 | ||
CJ175540.1 | - | 4 | 405 |