Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160096.1 | NP_001153568.1 | 2 | 463 | ||
NM_001160097.1 | NP_001153569.1 | 2 | 520 | |||
NM_001160098.1 | NP_001153570.1 | 2 | 463 | |||
NM_001160099.1 | NP_001153571.1 | 2 | 280 | |||
NM_021386.4 | NP_067361.2 | 2 | 280 | |||
NM_023878.3 | NP_076367.2 | 2 | 520 | |||
GenBank mRNA | AF124425.1 | - | 2 | 249 | ||
AK020131.1 | - | 2 | 484 | |||
BC021770.1 | - | 2 | 473 | 5040100 | ||
BC029019.1 | - | 273 | 5356256 | |||
CA467408.1 | - | 557 | ||||
CA481582.1 | - | 2 | 471 | |||
CK332009.1 | - | 2 | 85 | |||
CK794879.1 | - | 2 | 280 |