Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022017.3 | NP_071300.2 | 6 | 1034 | ||
GenBank mRNA | AB021875.2 | - | 5 | 961 | ||
AF208026.1 | - | 6 | 1002 | |||
AF263522.1 | - | 5 | 992 | |||
AF279672.1 | - | 6 | 1018 | |||
AJ296078.1 | - | 5 | 953 | |||
AK002735.1 | - | 6 | 1035 | |||
AK083223.1 | - | 6 | 1005 | |||
AK158652.1 | - | 2 | 887 | |||
BC127052.1 | - | 6 | 999 | 40109678 |