Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRPV4 SIRNA Assays ›
          Gene Symbol:
          Trpv4
          Gene Name:
          transient receptor potential cation channel, subfamily V, member 4
          Gene Aliases:
          0610033B08Rik, OTRPC4, Trp12, VR-OAC, VRL-2, VROAC
          Chromosome Location:
          Chr.5: 114622152 - 114658435 on Build GRCm38
          UniGene ID:
          Mm.266450
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s82062

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Trpv4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022017.3 NP_071300.2 10 1759
          GenBank mRNA AB021875.2 - 9 1686
          AF208026.1 - 10 1727
          AF263522.1 - 9 1717
          AF279672.1 - 10 1743
          AJ296078.1 - 9 1678
          AK083223.1 - 10 1730
          AK158652.1 - 6 1612
          BC127052.1 - 10 1724 40109678

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          response to osmotic stress
          hypotonic response
          actin filament organization
          cell-cell junction assembly
          positive regulation of cytosolic calcium ion concentration
          osmosensory signaling pathway
          negative regulation of neuron projection development
          vasopressin secretion
          positive regulation of microtubule depolymerization
          actin cytoskeleton reorganization
          cellular response to heat
          hyperosmotic salinity response
          cortical microtubule organization
          microtubule polymerization
          regulation of response to osmotic stress
          transmembrane transport
          calcium ion import
          calcium ion transmembrane transport
          cellular response to osmotic stress
          cellular hypotonic response
          nucleotide binding
          actin binding
          osmosensor activity
          protein kinase C binding
          ion channel activity
          cation channel activity
          calcium channel activity
          protein binding
          calmodulin binding
          ATP binding
          microtubule binding
          protein kinase binding
          SH2 domain binding
          alpha-tubulin binding
          beta-tubulin binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0