Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022018.3 | NP_071301.2 | 14 | 2804 | ||
GenBank mRNA | AB049355.1 | - | 14 | 2644 | ||
AK029847.1 | - | 10 | 2116 | |||
AK136597.1 | - | 14 | 2714 | |||
AK170193.1 | - | 15 | 2882 | |||
AK170347.1 | - | 5 | 1399 | |||
AK187603.1 | - | 1 | 102 | |||
AK191335.1 | - | 1 | 102 | |||
AK199589.1 | - | 1 | 99 | |||
AK209783.1 | - | 1 | 102 | |||
BC058234.1 | - | 12 | 2404 | 6306849 | ||
BC099690.1 | - | 14 | 2711 | |||
BC137843.1 | - | 10 | 2181 | 8860862 |