Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SIRT2 SIRNA Assays ›
          Gene Symbol:
          Sirt2
          Gene Name:
          sirtuin 2
          Gene Aliases:
          5730427M03Rik, SIR2L2, Sir2l
          Chromosome Location:
          Chr.7: 28766752 - 28788665 on Build GRCm38
          UniGene ID:
          Mm.272443
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s82218

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sirt2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122765.1 NP_001116237.1 13 978
          NM_001122766.1 NP_001116238.1 11 815
          NM_022432.4 NP_071877.3 14 1025
          GenBank mRNA AB221570.1 - 14 930
          AF299337.1 - 14 1010
          AK014042.1 - 13 949
          AK144431.1 - 13 922
          AK146412.1 - 13 955
          BC021439.1 - 14 1029 5039278

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein deacetylation
          autophagy
          cell cycle
          mitotic nuclear division
          nervous system development
          negative regulation of cell proliferation
          negative regulation of autophagy
          negative regulation of peptidyl-threonine phosphorylation
          phosphatidylinositol 3-kinase signaling
          histone deacetylation
          myelination in peripheral nervous system
          cell differentiation
          regulation of myelination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          cellular response to oxidative stress
          peptidyl-lysine deacetylation
          cellular response to hepatocyte growth factor stimulus
          negative regulation of protein catabolic process
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of DNA binding
          protein kinase B signaling
          cellular lipid catabolic process
          regulation of fat cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of meiotic nuclear division
          negative regulation of striated muscle tissue development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          hepatocyte growth factor receptor signaling pathway
          negative regulation of oligodendrocyte differentiation
          cell division
          meiotic cell cycle
          regulation of cell cycle
          positive regulation of cell division
          positive regulation of attachment of spindle microtubules to kinetochore
          negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
          cellular response to caloric restriction
          negative regulation of oligodendrocyte progenitor proliferation
          histone H3 deacetylation
          histone H4 deacetylation
          cellular response to molecule of bacterial origin
          cellular response to hypoxia
          cellular response to epinephrine stimulus
          tubulin deacetylation
          positive regulation of execution phase of apoptosis
          positive regulation of oocyte maturation
          negative regulation of NLRP3 inflammasome complex assembly
          negative regulation of defense response to bacterium
          ripoptosome assembly involved in necroptotic process
          negative regulation of reactive oxygen species metabolic process
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process involved in cellular response to hypoxia
          chromatin binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          zinc ion binding
          protein-N-terminal asparagine amidohydrolase activity
          UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
          hydrolase activity
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in linear amides
          iprodione amidohydrolase activity
          (3,5-dichlorophenylurea)acetate amidohydrolase activity
          protein deacetylase activity
          4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
          didemethylisoproturon amidohydrolase activity
          N-isopropylacetanilide amidohydrolase activity
          N-cyclohexylformamide amidohydrolase activity
          isonicotinic acid hydrazide hydrolase activity
          cis-aconitamide amidase activity
          gamma-N-formylaminovinylacetate hydrolase activity
          NAD-dependent protein deacetylase activity
          histone acetyltransferase binding
          histone deacetylase binding
          tubulin deacetylase activity
          ubiquitin binding
          N2-acetyl-L-lysine deacetylase activity
          O-succinylbenzoate synthase activity
          indoleacetamide hydrolase activity
          N-acetylcitrulline deacetylase activity
          metal ion binding
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          N-acetylgalactosamine-6-phosphate deacetylase activity
          beta-tubulin binding
          NAD binding
          diacetylchitobiose deacetylase activity
          chitooligosaccharide deacetylase activity
          NAD+ binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc