Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025375.2 | NP_001020546.1 | 6 | 649 | ||
NM_001025376.2 | NP_001020547.1 | 6 | 538 | |||
NM_023191.3 | NP_075680.1 | 6 | 543 | |||
GenBank mRNA | AF309554.1 | - | 6 | 501 | ||
AK010283.1 | - | 6 | 489 | |||
AK012342.1 | - | 6 | 555 | |||
AK013114.1 | - | 6 | 507 | |||
AK077037.1 | - | 583 | ||||
AK142498.1 | - | 6 | 506 | |||
AK153862.1 | - | 6 | 576 | |||
AK167706.1 | - | 6 | 576 | |||
BC023026.1 | - | 6 | 488 | 5374890 | ||
EF011623.1 | - | 6 | 486 |