Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025375.2 | NP_001020546.1 | 8, 9 | 873 | ||
NM_001025376.2 | NP_001020547.1 | 8, 9 | 762 | |||
NM_023191.3 | NP_075680.1 | 8, 9 | 767 | |||
GenBank mRNA | AF309554.1 | - | 8, 9 | 725 | ||
AK010283.1 | - | 8, 9 | 713 | |||
AK012342.1 | - | 8, 9 | 779 | |||
AK013114.1 | - | 8, 9 | 731 | |||
AK077037.1 | - | 807 | ||||
AK142498.1 | - | 8, 9 | 730 | |||
AK153862.1 | - | 8, 9 | 800 | |||
AK167706.1 | - | 8, 9 | 800 | |||
AK205840.1 | - | 2, 3 | 159 | |||
BC023026.1 | - | 8, 9 | 712 | 5374890 | ||
EF011623.1 | - | 8, 9 | 710 |