Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285429.1 | NP_001272358.1 | 6 | 552 | ||
NM_001285430.1 | NP_001272359.1 | 6 | 552 | |||
NM_001285431.1 | NP_001272360.1 | 5 | 480 | |||
NM_001285432.1 | NP_001272361.1 | 5 | 483 | |||
NM_001285433.1 | NP_001272362.1 | 5 | 480 | |||
NM_001285434.1 | NP_001272363.1 | 6 | 537 | |||
NM_023240.3 | NP_075729.2 | 6 | 555 | |||
NM_029663.2 | NP_083939.1 | 8 | 1700 | |||
GenBank mRNA | AF304351.1 | - | 6 | 518 | ||
AK010308.1 | - | 2 | 171 | |||
AK010347.1 | - | 7 | 530 | |||
AK017616.1 | - | 5 | 435 | |||
AK017796.1 | - | 6 | 518 | |||
AK044934.1 | - | 6 | 507 | |||
BC013059.1 | - | 6 | 485 | |||
BC079855.1 | - | 8 | 1646 | 6403006 | ||
BC082536.1 | - | 8 | 1631 | |||
BQ177630.1 | - | 6 | 522 | |||
DV062370.1 | - | 5 | 487 |