Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025739.2 | NP_080015.3 | 13 | 1827 | ||
GenBank mRNA | AK016436.1 | - | 12 | 1075 | ||
AK172415.1 | - | 9 | 894 | |||
BC034875.1 | - | 8 | 778 | |||
BC056359.1 | - | 13 | 1827 | 6813564 | ||
BC058415.1 | - | 13 | 1827 | 6813564 |