Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025824.3 | NP_080100.1 | 10 | 1131 | ||
GenBank mRNA | AK004784.2 | - | 10 | 1131 | ||
AK008059.1 | - | 2 | 224 | |||
AK013980.1 | - | 10 | 1125 | |||
AK029030.1 | - | 10 | 1143 | |||
AK076086.1 | - | 10 | 1132 | |||
AK076560.1 | - | 10 | 1132 | |||
AK088322.1 | - | 10 | 1139 | |||
AK151919.1 | - | 10 | 1119 | |||
AK152939.1 | - | 10 | 1118 | |||
AK153538.1 | - | 10 | 1120 | |||
AK161192.1 | - | 10 | 1129 | |||
AK166247.1 | - | 10 | 1136 | |||
AK167056.1 | - | 10 | 1118 | |||
AK167074.1 | - | 10 | 1103 | |||
AK167514.1 | - | 10 | 1090 | |||
AK167982.1 | - | 10 | 1137 | |||
AK168158.1 | - | 10 | 1143 | |||
AK191609.1 | - | 3 | 334 | |||
AK198435.1 | - | 3 | 334 | |||
AK200532.1 | - | 3 | 334 | |||
AK204270.1 | - | 3 | 336 | |||
BC005466.1 | - | 10 | 1140 | 3601190 | ||
BC028865.1 | - | 10 | 1119 | 4910833 |