Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025824.3 | NP_080100.1 | 8 | 773 | ||
GenBank mRNA | AK004784.2 | - | 8 | 773 | ||
AK013980.1 | - | 8 | 767 | |||
AK029030.1 | - | 8 | 784 | |||
AK076086.1 | - | 8 | 774 | |||
AK076560.1 | - | 8 | 774 | |||
AK087980.1 | - | 8 | 761 | |||
AK088322.1 | - | 8 | 781 | |||
AK133973.1 | - | 8 | 774 | |||
AK151919.1 | - | 8 | 761 | |||
AK152939.1 | - | 8 | 760 | |||
AK153538.1 | - | 8 | 762 | |||
AK161192.1 | - | 8 | 771 | |||
AK166247.1 | - | 8 | 778 | |||
AK167056.1 | - | 8 | 758 | |||
AK167074.1 | - | 8 | 745 | |||
AK167514.1 | - | 8 | 732 | |||
AK167982.1 | - | 8 | 779 | |||
AK168158.1 | - | 8 | 785 | |||
AK198677.1 | - | 5 | 389 | |||
AK215707.1 | - | 4 | 361 | |||
BC005466.1 | - | 8 | 782 | 3601190 | ||
BC028865.1 | - | 8 | 761 | 4910833 |