Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284332.1 | NP_001271261.1 | 4 | 534 | ||
NM_025858.3 | NP_080134.1 | 4 | 531 | |||
NM_026381.5 | NP_080657.1 | 2 | 231 | |||
GenBank mRNA | AF520699.1 | - | 4 | 513 | ||
AK009225.1 | - | 4 | 471 | |||
AK018291.1 | - | 2 | 187 | |||
AK020877.1 | - | 2 | 208 | |||
AK033545.1 | - | 4 | 491 | |||
AK045368.1 | - | 4 | 461 | |||
AK081060.1 | - | 2 | 209 | |||
AK088188.1 | - | 3 | 227 | |||
AK090357.1 | - | 2 | 385 | |||
AK090373.1 | - | 4 | 470 | |||
AK150460.1 | - | 4 | 496 | |||
AK151632.1 | - | 4 | 496 | |||
AK205406.1 | - | 1 | 103 | |||
AK212074.1 | - | 2 | 190 | |||
BC010238.1 | - | 4 | 495 | 3581921 | ||
BC017600.1 | - | 2 | 143 | 4487600 | ||
BC057089.1 | - | 2 | 183 | 5689110 |