Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024185.4 | NP_077147.2 | 14 | 1372 | ||
GenBank mRNA | AK002900.1 | - | 14 | 1323 | ||
AK009891.1 | - | 14 | 1351 | |||
AK010050.1 | - | 14 | 1338 | |||
AK049869.1 | - | 13 | 1296 | |||
AK193641.1 | - | 3 | 218 | |||
BC002225.1 | - | 14 | 1354 | |||
BC027202.1 | - | 14 | 1276 | 4217391 |