Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam103a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025997.2 | NP_080273.1 | 2 | 173 | ||
GenBank mRNA | AK010694.1 | - | 2 | 129 | ||
AK011881.1 | - | 2 | 173 | |||
AK018614.1 | - | 2 | 172 | |||
AK079819.1 | - | 2 | 152 | |||
AK081301.1 | - | 2 | 165 | |||
AK085230.1 | - | 2 | 171 | |||
AK150938.1 | - | 2 | 140 | |||
AK160347.1 | - | 2 | 172 | |||
AK169392.1 | - | 2 | 202 | |||
BC058367.1 | - | 2 | 96 | 6830048 | ||
BC091760.1 | - | 2 | 133 | 5042613 | ||
BC096399.1 | - | 2 | 131 | 876882 |