Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam120b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024203.3 | NP_077165.2 | 2 | 1468 | ||
GenBank mRNA | AK016568.1 | - | 2 | 1434 | ||
AK028911.1 | - | 2 | 1398 | |||
AK050319.1 | - | 1 | 1183 | |||
AK076185.1 | - | 2 | 1434 | |||
AK087438.1 | - | 2 | 1415 | |||
AK132211.1 | - | 2 | 1446 | |||
AK220425.1 | - | 2 | 1430 | |||
BC003200.1 | - | 1 | 371 | |||
BC066088.1 | - | 2 | 1513 | |||
BC085163.1 | - | 2 | 1466 | 30618952 | ||
DQ873694.1 | - | 2 | 1449 |