Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042499.1 | NP_001035964.1 | 5 | 465 | ||
GenBank mRNA | AK016099.1 | - | 5 | 474 | ||
AK028195.1 | - | 5 | 459 | |||
AK028269.1 | - | 5 | 452 | |||
AK156982.1 | - | 4 | 3964 | |||
BC050194.1 | - | 5 | 446 | 30022531 |