Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290761.1 | NP_001277690.1 | 5 | 1224 | ||
NM_001290762.1 | NP_001277691.1 | 5 | 1301 | |||
NM_026393.2 | NP_080669.1 | 6 | 1254 | |||
GenBank mRNA | AA166262.1 | - | 1 | 290 | ||
AK012340.1 | - | 5 | 1057 | |||
AK146521.1 | - | 980 | ||||
BC030039.1 | - | 6 | 957 | 1313824 |