Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205286.1 | NP_001192215.1 | 8 | 1296 | ||
NM_001205287.1 | NP_001192216.1 | 8 | 1344 | |||
NM_026407.3 | NP_080683.2 | 8 | 1548 | |||
GenBank mRNA | AK011646.1 | - | 8 | 1525 | ||
AK017817.1 | - | 8 | 1506 | |||
AK133638.1 | - | 8 | 1520 | |||
AK160576.1 | - | 8 | 1297 | |||
AK169127.1 | - | 8 | 1346 | |||
BC020318.2 | - | 8 | 1503 | 5250532 |