Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026417.4 | NP_080693.2 | 4 | 654 | ||
GenBank mRNA | AK009618.1 | - | 4 | 640 | ||
AK031602.1 | - | 1 | 1233 | |||
AK083301.1 | - | 4 | 634 | |||
AK159227.1 | - | 4 | 615 | |||
AK166347.1 | - | 4 | 642 | |||
BC003936.1 | - | 5 | 829 | 3486503 |