Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sympk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026605.2 | NP_080881.2 | 18 | 2521 | ||
GenBank mRNA | AK019497.1 | - | 13 | 1997 | ||
AK146894.1 | - | 18 | 2502 | |||
AK155093.1 | - | 18 | 2522 | |||
AK155375.1 | - | 18 | 2476 | |||
AK165591.1 | - | 18 | 2500 | |||
AK168541.1 | - | 18 | 2502 | |||
AK168757.1 | - | 18 | 2502 | |||
AK172040.1 | - | 19 | 2550 | |||
AK179712.1 | - | 3 | 230 | |||
AK183486.1 | - | 1 | 36 | |||
AK207325.1 | - | 1 | 36 | |||
BC049852.1 | - | 18 | 2466 | 4163760 |