Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039188.1 | NP_001034277.1 | 6 | 1002 | ||
NM_001177868.1 | NP_001171339.1 | 6 | 727 | |||
NM_001177869.1 | NP_001171340.1 | 5 | 663 | |||
NM_026830.2 | NP_081106.1 | 6 | 970 | |||
GenBank mRNA | AK047047.1 | - | 6 | 1921 | ||
AK047251.1 | - | 6 | 1923 | |||
AK147260.1 | - | 6 | 2125 | |||
AK147653.1 | - | 6 | 2125 | |||
AK154980.1 | - | 6 | 970 | |||
AK170652.1 | - | 6 | 1002 | |||
AK171375.1 | - | 6 | 707 | |||
AY946044.1 | - | 3 | 354 | |||
BC080680.1 | - | 6 | 816 | 6330949 |