Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PINK1 SIRNA Assays ›
          Gene Symbol:
          Pink1
          Gene Name:
          PTEN induced putative kinase 1
          Gene Aliases:
          1190006F07Rik, AU042772, AW557854, BRPK, mFLJ00387
          Chromosome Location:
          Chr.4: 138313409 - 138326296 on Build GRCm38
          UniGene ID:
          Mm.18539
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s87251

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pink1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_026880.2 NP_081156.2 8 1592
          GenBank mRNA AB053476.1 - 8 1524
          AF316872.1 - 8 1570
          AK004507.1 - 4 440
          AK050322.1 - 8 1572
          AK165480.1 - 8 1570
          AK181855.1 - 2 129
          AK220318.1 - 8 1546
          BC018561.1 - 8 1416
          BC044743.1 - 3 288
          BC067066.1 - 8 1560 30542123
          BC069968.1 - 9 1450

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy
          positive regulation of protein phosphorylation
          regulation of oxidative phosphorylation
          protein phosphorylation
          autophagy
          response to stress
          response to oxidative stress
          mitochondrion organization
          regulation of hydrogen peroxide metabolic process
          negative regulation of gene expression
          regulation of mitochondrion organization
          positive regulation of macroautophagy
          negative regulation of macroautophagy
          phosphorylation
          protein ubiquitination
          peptidyl-serine phosphorylation
          respiratory electron transport chain
          regulation of protein ubiquitination
          positive regulation of synaptic transmission, dopaminergic
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of dopamine secretion
          positive regulation of catecholamine secretion
          cellular response to oxidative stress
          positive regulation of protein dephosphorylation
          intracellular signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of protein complex assembly
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of translation
          protein stabilization
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of mitochondrial membrane potential
          cellular response to hypoxia
          positive regulation of mitochondrial fission
          positive regulation of release of cytochrome c from mitochondria
          negative regulation of mitochondrial fission
          cellular response to toxic substance
          mitophagy in response to mitochondrial depolarization
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          negative regulation of autophagosome assembly
          positive regulation of mitochondrial electron transport, NADH to ubiquinone
          negative regulation of mitophagy
          negative regulation of oxidative stress-induced cell death
          regulation of protein targeting to mitochondrion
          negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
          negative regulation of hydrogen peroxide-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of cristae formation
          positive regulation of protein targeting to mitochondrion
          regulation of reactive oxygen species metabolic process
          negative regulation of reactive oxygen species metabolic process
          nucleotide binding
          magnesium ion binding
          protease binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          calcium-dependent protein kinase activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          ubiquitin protein ligase binding
          protein kinase B binding
          metal ion binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix