Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTAGE5 SIRNA Assays ›
Gene Symbol:
CTAGE5
Gene Name:
CTAGE family member 5, ER export factor
Gene Aliases:
MEA6, MGEA, MGEA11, MGEA6
Chromosome Location:
Chr.14: 39265264 - 39351193 on Build GRCh38
UniGene ID:
Hs.509200
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8745
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTAGE5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001247988.1
NP_001234917.1
18
1649
NM_001247989.1
NP_001234918.1
18
1833
NM_001247990.1
NP_001234919.1
18
1723
NM_005930.3
NP_005921.2
18
1818
NM_203354.2
NP_976229.1
18
1569
NM_203355.2
NP_976230.1
18
1818
NM_203356.2
NP_976231.1
18
1649
XM_005267648.4
XP_005267705.1
18
1529
XM_006720148.2
XP_006720211.1
18
1531
XM_011536775.2
XP_011535077.1
19
1896
XM_011536778.2
XP_011535080.1
19
1584
XM_011536784.2
XP_011535086.1
18
1447
XM_011536785.2
XP_011535087.1
18
1439
XM_017021314.1
XP_016876803.1
19
1912
XM_017021315.1
XP_016876804.1
19
1613
XM_017021316.1
XP_016876805.1
19
1600
XM_017021317.1
XP_016876806.1
19
2021
XM_017021318.1
XP_016876807.1
19
1642
XM_017021319.1
XP_016876808.1
19
1578
XM_017021320.1
XP_016876809.1
18
1604
XM_017021321.1
XP_016876810.1
19
1897
XM_017021322.1
XP_016876811.1
18
1577
XM_017021323.1
XP_016876812.1
19
1396
XM_017021324.1
XP_016876813.1
19
1484
XM_017021325.1
XP_016876814.1
18
1537
XM_017021326.1
XP_016876815.1
18
1447
XM_017021327.1
XP_016876816.1
18
1439
XM_017021328.1
XP_016876817.1
18
1440
XM_017021329.1
XP_016876818.1
18
1419
XM_017021330.1
XP_016876819.1
19
1521
GenBank mRNA
AB209528.1
-
17
2127
AF338233.1
-
18
1663
AF338234.1
-
18
1696
AK091252.1
-
18
2027
AK298935.1
-
17
1472
AK315254.1
-
18
1539
AL832233.1
-
17
1510
AL833415.1
-
17
1400
BC039017.1
-
18
1611
BC051363.1
-
18
1436
4576721
BC064355.1
-
18
1779
4803802
BX640994.1
-
18
1551
BX648014.1
-
18
1551
U73682.1
-
18
1528
U94780.1
-
18
1796
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
cargo loading into vesicle
positive regulation of catalytic activity
protein localization to endoplasmic reticulum exit site
regulation of guanyl-nucleotide exchange factor activity
protein binding
enzyme activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0