Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175160.3 | NP_780369.1 | 6 | 800 | ||
GenBank mRNA | AK014489.1 | - | 1 | 1668 | ||
AK038593.1 | - | 6 | 802 | |||
BC026570.1 | - | 2 | 381 | 4984252 | ||
BC119378.1 | - | 3 | 539 | 8734127 | ||
BC144878.1 | - | 3 | 539 | 9053426 |