Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 17 | 2778 | ||
NM_001252579.1 | NP_001239508.1 | 17 | 2624 | |||
NM_028072.5 | NP_082348.2 | 17 | 2620 | |||
GenBank mRNA | AK008108.1 | - | 8 | 883 | ||
AK028874.1 | - | 17 | 2535 | |||
AK034712.1 | - | 17 | 2605 | |||
AK036685.1 | - | 17 | 2556 | |||
AK049170.1 | - | 12 | 1535 | |||
AK081643.1 | - | 17 | 2578 | |||
AK129316.1 | - | 17 | 2778 | |||
AK133336.1 | - | 2594 | ||||
AK165183.1 | - | 17 | 2532 | |||
AY101177.1 | - | 16 | 2439 | |||
BC027238.1 | - | 6 | 594 | |||
BC062900.1 | - | 17 | 2576 | |||
BC141086.1 | - | 16 | 2373 | 9055698 |