Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164798.1 | NP_001158270.1 | 10 | 1367 | ||
NM_001164799.1 | NP_001158271.1 | 10 | 1367 | |||
NM_001164800.1 | NP_001158272.1 | 10 | 1367 | |||
NM_001164801.1 | NP_001158273.1 | 10 | 1367 | |||
NM_001164802.1 | NP_001158274.1 | 10 | 1367 | |||
NM_024249.5 | NP_077211.4 | 10 | 1367 | |||
GenBank mRNA | AK007977.1 | - | 10 | 1257 | ||
AK032245.1 | - | 10 | 1291 | |||
AK090050.1 | - | 10 | 1343 | |||
AK159948.1 | - | 10 | 1329 | |||
AK165672.1 | - | 10 | 1332 | |||
AK180057.1 | - | 3 | 247 | |||
AK185748.1 | - | 2 | 138 | |||
AK189102.1 | - | 3 | 232 | |||
BC043299.1 | - | 1 | 64 | |||
BC082300.1 | - | 10 | 1338 | 30362204 | ||
BC088811.1 | - | 10 | 1303 | 6493546 |