Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185002.1 | NP_001171931.1 | 1 | 969 | ||
GenBank mRNA | AK007253.1 | - | 1 | 111 |