Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610037L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028754.2 | NP_083030.1 | 6 | 608 | ||
GenBank mRNA | AK002776.1 | - | 6 | 608 | ||
AK003575.1 | - | 7 | 423 | |||
AK032590.1 | - | 399 | ||||
AK050486.1 | - | 7 | 442 | |||
AK050533.1 | - | 7 | 405 | |||
AK160674.1 | - | 7 | 412 | |||
BC019215.1 | - | 6 | 614 | 5065090 | ||
BC037649.1 | - | 6 | 408 |