Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102436.1 | NP_001095906.1 | 8 | 991 | ||
NM_001102437.1 | NP_001095907.1 | 8 | 1118 | |||
NM_001102438.1 | NP_001095908.1 | 8 | 1118 | |||
NM_028793.3 | NP_083069.1 | 8 | 991 | |||
GenBank mRNA | AK005001.2 | - | 7 | 907 | ||
AK050450.1 | - | 8 | 957 | |||
AK147839.1 | - | 8 | 1118 | |||
BC035202.1 | - | 8 | 1047 | |||
BC053518.1 | - | 8 | 1085 | |||
BC061484.1 | - | 7 | 889 | 5355466 | ||
BC084584.1 | - | 8 | 923 | 5257773 | ||
BC090641.1 | - | 8 | 942 |