Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm21379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293057.1 | NP_001279986.1 | 1 | 1126 |
Gene Symbol: Rbm31y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028970.1 | NP_083246.1 | 1 | 1126 | ||
GenBank mRNA | AK017055.1 | - | 1 | 1129 |