Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 8430419L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028982.4 | NP_083258.2 | 10 | 1546 | ||
GenBank mRNA | AK018428.1 | - | 10 | 1490 | ||
AK039401.1 | - | 10 | 1482 | |||
AK155128.1 | - | 10 | 1491 | |||
AK220395.1 | - | 9 | 1196 | |||
BC003918.1 | - | 2 | 145 | |||
BC038002.1 | - | 9 | 1207 | |||
BC079676.1 | - | 10 | 1473 | |||
BC137674.1 | - | 10 | 1525 | 8860693 | ||
BC137676.1 | - | 10 | 1525 | 8860695 |