Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081293.1 | NP_001074762.1 | 4 | 759 | ||
GenBank mRNA | AK012860.1 | - | 3 | 1466 | ||
AK015972.1 | - | 4 | 616 | |||
AK083674.1 | - | 4 | 528 | |||
AK087157.1 | - | 4 | 531 | |||
BC067054.1 | - | 4 | 564 | 30542928 |