Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165980.1 | NP_001159452.1 | 10 | 1279 | ||
NM_001165981.1 | NP_001159453.1 | 10 | 1279 | |||
NM_198005.2 | NP_932122.2 | 10 | 1279 | |||
GenBank mRNA | AK014717.1 | - | 10 | 1243 | ||
AK079462.1 | - | 10 | 1073 | |||
AK081839.1 | - | 10 | 1278 | |||
AK160679.1 | - | 10 | 1246 | |||
BC031546.1 | - | 10 | 1171 | 3669736 |