Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253681.1 | NP_001240610.1 | 14 | 1757 | ||
NM_001253686.1 | NP_001240615.1 | 13 | 1742 | |||
NM_001253687.1 | NP_001240616.1 | 12 | 1646 | |||
NM_001253688.1 | NP_001240617.1 | 12 | 1556 | |||
NM_029673.3 | NP_083949.2 | 14 | 1790 | |||
GenBank mRNA | AK006977.1 | - | 2 | 135 | ||
AK033126.1 | - | 12 | 1557 | |||
AK037930.1 | - | 1 | 2876 | |||
AK038129.1 | - | 14 | 1780 | |||
AK049189.1 | - | 14 | 1701 | |||
AK152588.1 | - | 13 | 1688 | |||
AK159909.1 | - | 13 | 1693 | |||
AK163491.1 | - | 14 | 1665 | |||
AK169365.1 | - | 12 | 1459 | |||
BC025034.1 | - | 2 | 535 | |||
BC055840.1 | - | 2 | 219 | |||
BC061010.1 | - | 3 | 261 | |||
BC080790.1 | - | 1 | 107 | |||
BC108356.1 | - | 6 | 688 | |||
BC145704.1 | - | 1 | 70 | |||
BC145706.1 | - | 1 | 70 |